N,N'''-1,6-Hexanediylbis(N'-cyanoguanidine) 98%

Supplier: Ambeed
Warning

A412115-25G A412115-100G A412115-10G
103838-288EA 15 USD
103838-288 76841-830 77115-846 77686-140
N,N'''-1,6-Hexanediylbis(N'-cyanoguanidine) 98%
N,N'''-1,6-Hexanediylbis(N'-cyanoguanidine)

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₈
Boiling Pt: 437.7 °C (760 mm Hg)
Flash Pt: 218.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00128128
CAS Number: 15894-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR