N,N',N''-Triphenyl-1,3,5-benzenetriamine 95%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tris(phenylamino)benzene

A277707-250MG A277707-1G
103813-362EA 29 USD
103813-362 103853-298 77099-120 77305-266
N,N',N''-Triphenyl-1,3,5-benzenetriamine 95%
N,N',N''-Triphenyl-1,3,5-benzenetriamine

Precaución: for Research Use Only

Formula: C₂₄H₂₁N₃
MW: 351.45 g/mol
Boiling Pt: 530.6 °C (760 mm Hg)
Melting Pt: 198 °C
Flash Pt: 297.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01463903
CAS Number: 102664-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR