Decyltrimethylammonium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N,N-Trimethyldecan-1-aminium chloride
A111030-100G
A111030-10G
A111030-25G
A111030-500G
A111030-5G
103808-058EA
324
USD
103808-058
103823-586
76687-576
76687-578
76687-580
76687-582
77284-076
Decyltrimethylammonium chloride 97%
Decyltrimethylammonium chloride
Precaución: for Research Use Only
Formula:
C₁₃H₃₀ClN MW: 235.8400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059967 CAS Number: 10108-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...