N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% in water

Supplier: Ambeed
Danger

A543238-100G A543238-10G A543238-500G A543238-5G
103839-770EA 5 USD
103839-770 76843-824 77017-590 77017-592 77017-594 77307-776
N,N,N-Trimethyl-1-adamantylammonium hydroxide 25% in water
N,N,N-Trimethyl-1-adamantylammonium hydroxide

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO
Storage Temperature: Refrigerator
MDL Number: MFCD27920795
CAS Number: 53075-09-5
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR