N,N'-(Oxybis(4,1-phenylene))diacetamide 97%

Supplier: Ambeed
Warning

A911647-1G A911647-100MG A911647-250MG
103841-370EA 625 USD
103841-370 76845-646 77244-762 77244-764
N,N'-(Oxybis(4,1-phenylene))diacetamide 97%
N,N'-(Oxybis(4,1-phenylene))diacetamide

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₃
MW: 284.3 g/mol
Boiling Pt: 554.4 °C (760 mm Hg)
Melting Pt: 228…229 °C at N/A mmHg
Flash Pt: 289.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00050458
CAS Number: 3070-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR