N,N'-(Hexane-1,6-diyl)bis(N-(2,2,6,6-tetramethylpiperidin-4-yl)formamide) 97%

Supplier: Ambeed
Warning

A124269-100G A124269-25G A124269-500G A124269-1000G
103808-292EA 113 USD
103808-292 77246-234 77246-236 77246-238 77312-646
N,N'-(Hexane-1,6-diyl)bis(N-(2,2,6,6-tetramethylpiperidin-4-yl)formamide) 97%
N,N'-(Hexane-1,6-diyl)bis(N-(2,2,6,6-tetramethylpiperidin-4-yl)formamide)

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂
Boiling Pt: 596.0±50.0 °C (760 mm Hg)
Melting Pt: 155 °C
Flash Pt: 314.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD25292833
CAS Number: 124172-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR