N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide] 97%

Supplier: Ambeed
Warning

A244037-100G A244037-10G A244037-500G A244037-5G A244037-1G A244037-25G A244037-1000G
103811-564EA 5 USD
103811-564 103851-388 77000-506 77000-508 77000-510 77090-594 77304-298 77304-300 77568-372
N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide] 97%
N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide]

Precaución: For research use only.

Formula: C₄₀H₆₄N₂O₄
Boiling Pt: 740.1 °C (760 mm Hg)
Melting Pt: 156…161 °C
Flash Pt: 401.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00134697
CAS Number: 23128-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR