N,6-dimethylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A780967-100MG A780967-250MG
103840-978EA 375 USD
103840-978 77171-230 77171-232
N,6-dimethylpyridin-2-amine 95%
N,6-dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697830
CAS Number: 97986-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR