tert-Butyl (cyanomethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-2-aminoacetonitrile

A162640-100G A162640-10G A162640-250MG A162640-1G A162640-25G A162640-5G
103805-644EA 6 USD
103805-644 103843-894 76720-180 76720-182 76720-184 77319-958 77319-960 77319-962
tert-Butyl (cyanomethyl)carbamate 95%
tert-Butyl (cyanomethyl)carbamate

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O₂
MW: 156.19 g/mol
Boiling Pt: 186 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00239390
CAS Number: 85363-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR