N-(Chloroacetoxy)succinimide 96%

Supplier: Ambeed
Danger

A112885-100MG A112885-10G A112885-250MG A112885-1G A112885-5G
103798-126EA 45 USD
103798-126 103823-962 77148-782 77148-784 77148-786 77310-792 77310-794
N-(Chloroacetoxy)succinimide 96%
N-(Chloroacetoxy)succinimide

Precaución: for Research Use Only

Formula: C₆H₆ClNO₄
MW: 191.57 g/mol
Boiling Pt: 277.3±42.0 °C (760 mm Hg)
Melting Pt: 109…110 °C
Flash Pt: 121.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044492
CAS Number: 27243-15-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR