N-(Benzyloxycarbonyloxy)succinimide 97%

Supplier: Ambeed
Warning

Synonyms: CBZ-OSu (Z-OSu)

A150028-100G A150028-500G A150028-25G A150028-1000G
103842-246EA 5 USD
103842-246 76719-686 76719-688 77317-758 77600-122
N-(Benzyloxycarbonyloxy)succinimide 97%
N-(Benzyloxycarbonyloxy)succinimide

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₅
MW: 249.22 g/mol
Melting Pt: 80…82 °C
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005513
CAS Number: 13139-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR