N-(Allyloxycarbonyloxy)succinimide 96%

Supplier: Ambeed
Warning

A795256-25G A795256-100G A795256-10G A795256-500G A795256-5G
103841-036EA 15 USD
103841-036 76845-280 77171-332 77171-334 77171-336 77308-346
N-(Allyloxycarbonyloxy)succinimide 96%
N-(Allyloxycarbonyloxy)succinimide

Precaución: for Research Use Only

Formula: C₈H₉NO₅
Boiling Pt: 269.471 °C (760 mm Hg)
Melting Pt: 21 °C at N/A mmHg
Flash Pt: 116.772 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684388
CAS Number: 135544-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR