Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%

Supplier: Ambeed
Danger

Synonyms: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

A190909-25G A190909-10G A190909-100G A190909-500G
103809-250EA 117 USD
103809-250 103846-938 77084-768 77287-478 77550-220 77748-220
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)

Precaución: for Research Use Only

Formula: C₁₉H₁₅NO₅
MW: 337.3 g/mol
Melting Pt: 150…153 °C
Flash Pt: 252.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010733
CAS Number: 82911-69-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR