1-Acetamido-7-hydroxynaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: N-(7-Hydroxynaphthalen-1-yl)acetamide

A216291-10G A216291-1G A216291-5G
103849-270EA 7 USD
103849-270 76721-832 76721-834 77087-844
1-Acetamido-7-hydroxynaphthalene 97%
1-Acetamido-7-hydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MW: 201.225 g/mol
Boiling Pt: 486.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00035710
CAS Number: 6470-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR