N-(6-Oxo-6,7-dihydro-1H-purin-2-yl)isobutyramide 98%

Supplier: Ambeed
Warning

A160222-250MG A160222-1G A160222-100G
103805-350EA 9 USD
103805-350 103843-580 77028-610 77319-500 77567-336
N-(6-Oxo-6,7-dihydro-1H-purin-2-yl)isobutyramide 98%
N-(6-Oxo-6,7-dihydro-1H-purin-2-yl)isobutyramide

Precaución: For research use only.

Formula: C₉H₁₁N₅O₂
MW: 221.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16038625
CAS Number: 21047-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR