N-(5-Fluoro-2-iodophenyl)acetamide 95%
Supplier: Ambeed
|
Warning
|
A102201-100MG
A102201-250MG
103796-336EA
41
USD
103796-336
103822-030
77093-558
77093-560
N-(5-Fluoro-2-iodophenyl)acetamide 95%
N-(5-Fluoro-2-iodophenyl)acetamide
Precaución: for Research Use Only
Formula:
C₈H₇FINO Melting Pt: 130…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD28405559 CAS Number: 1173707-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...