N-(5-Bromopentyl)phthalimide 97%
Supplier: Ambeed
|
Warning
|
A360468-10G
A360468-250MG
A360468-25G
A360468-5G
A360468-1G
103826-256EA
13
USD
103826-256
103837-266
76724-588
76724-590
76840-476
76840-478
77306-710
N-(5-Bromopentyl)phthalimide 97%
N-(5-Bromopentyl)phthalimide
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BrNO₂ MW: 296.16 g/mol Melting Pt: 59…64 °C Flash Pt: 191.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060522 CAS Number: 954-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...