N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(3-pyridyl)-2-pyrimidineamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine, N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine
A127173-10G
A127173-1G
A127173-25G
A127173-5G
103800-926EA
5
USD
103800-926
77271-834
77313-168
77313-170
77313-172
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine 98%
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine
Precaución: for Research Use Only
Formula:
C₁₆H₁₅N₅ Boiling Pt: 537.3 °C (760 mm Hg) Melting Pt: 133…135 °C Flash Pt: 278.765 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09028125 CAS Number: 152460-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...