4'-Methoxyacetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Methoxyphenyl)acetamide

A921289-25G A921289-100G
103841-406EA 9 USD
103841-406 76845-672 77294-240
4'-Methoxyacetanilide 98%
4'-Methoxyacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 345.5 °C (760 mm Hg)
Melting Pt: 129…132 °C
Flash Pt: 162.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00014963
CAS Number: 51-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR