4-Acetamidocyclohexanol (cis- and trans- mixture) 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-Hydroxycyclohexyl)acetamide(cis- and trans- mixture)

A231643-100G A231643-10G A231643-500G A231643-1000G A231643-25G A231643-5G
103809-636EA 218 USD
103809-636 103850-538 77288-404 77288-406 77288-408 77303-918 77303-920 77303-922
4-Acetamidocyclohexanol (cis- and trans- mixture) 97%
4-Acetamidocyclohexanol (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
Boiling Pt: 357.2 °C (760 mm Hg)
Melting Pt: 162 °C
Flash Pt: 169.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06797038
CAS Number: 23363-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR