N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 97%

Supplier: Ambeed
Warning

A311945-100G A311945-10G A311945-1G A311945-5G
103814-686EA 6 USD
103814-686 103835-876 76723-834 76723-836 76838-686 76838-688
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 97%
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClN₃O₂
MW: 289.72 g/mol
Boiling Pt: 518.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09263704
CAS Number: 848133-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR