N-(4-Bromopyridin-2-yl)acetamide 95%

Supplier: Ambeed
Warning

A270233-250MG A270233-1G A270233-5G
103812-994EA 41 USD
103812-994 103852-900 76827-314 77092-668 77092-670
N-(4-Bromopyridin-2-yl)acetamide 95%
N-(4-Bromopyridin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11976392
CAS Number: 1026796-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR