N-(4-Bromophenyl)-4-phenylthiazol-2-amine 97%
Supplier: Ambeed
|
Warning
|
A1006566-1MG
A1006566-10MG
A1006566-5MG
103791-382EA
127
USD
103791-382
103791-876
103791-878
103799-224
76646-684
77296-790
77296-792
N-(4-Bromophenyl)-4-phenylthiazol-2-amine 97%
N-(4-Bromophenyl)-4-phenylthiazol-2-amine
Precaución: for Research Use Only
Formula:
C₁₅H₁₁BrN₂S MW: 331.23 g/mol Storage Temperature: Refrigerator |
CAS Number:
108237-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...