N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide 97%

Supplier: Ambeed
Warning

A152370-100G A152370-10G A152370-1G A152370-25G A152370-5G
103804-382EA 6 USD
103804-382 103842-550 76871-658 76871-660 77318-162 77318-164 77318-166
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide 97%
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₄O
MW: 262.351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12457658
CAS Number: 262368-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR