Amsacrine 98%

Supplier: Ambeed
Warning

Synonyms: m-AMSA, N-[4-(acridin-9-ylamino)-3-methoxyphenyl]methanesulphonamide, Amsidine

A472083-10MG A472083-1MG
103791-680EA 63 USD
103791-680 103792-730 103799-656 103828-038 76843-026 77241-246
Amsacrine 98%
Amsacrine

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₃O₃S
MW: 393.46 g/mol
Boiling Pt: 563 °C (760 mm Hg)
Melting Pt: 230…240 °C
Flash Pt: 294.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00242748
CAS Number: 51264-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR