4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulphonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulphonamide

A197110-100G A197110-5G
103847-508EA 24 USD
103847-508 77030-022 77085-494
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde 97%
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₈FN₃O₃S
MW: 351.40 g/mol
Boiling Pt: 536.594 °C (760 mm Hg)
Flash Pt: 278.322 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458342
CAS Number: 147118-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR