N-(3-Cyanophenyl)benzamide 95%

Supplier: Ambeed
Warning

A134261-100MG A134261-250MG A134261-1G
103801-994EA 157 USD
103801-994 76972-268 76972-270 77298-706
N-(3-Cyanophenyl)benzamide 95%
N-(3-Cyanophenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00451474
CAS Number: 141990-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR