3'-Chloro-4'-hydroxyacetanilide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-hydroxyacetanilide, N-(3-Chloro-4-hydroxyphenyl)acetamide

A101390-10G A101390-250MG A101390-1G A101390-5G
103795-952EA 30 USD
103795-952 103821-636 77233-084 77233-086 77297-086 77297-088
3'-Chloro-4'-hydroxyacetanilide 95%
3'-Chloro-4'-hydroxyacetanilide

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
Boiling Pt: 379.9 °C (760 mm Hg)
Melting Pt: 143…144 °C
Flash Pt: 183.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318123
CAS Number: 3964-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR