N-(3-Bromopropyl)phthalimide 97%

Supplier: Ambeed
Warning

A101368-100G A101368-10G A101368-25G A101368-500G A101368-5G
103821-618EA 6 USD
103821-618 76970-152 76970-154 76970-156 76970-158 77297-070
N-(3-Bromopropyl)phthalimide 97%
N-(3-Bromopropyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 361.4 °C (760 mm Hg)
Melting Pt: 72…74 °C
Flash Pt: 172.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005904
CAS Number: 5460-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR