5'-Acetamido-2'-hydroxyacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: N-(3-Acetyl-4-hydroxyphenyl)acetamide

A231941-100G A231941-10G A231941-1G A231941-25G A231941-5G
103810-804EA 7 USD
103810-804 103850-558 76722-256 76722-258 77089-468 77089-470 77089-472
5'-Acetamido-2'-hydroxyacetophenone 97%
5'-Acetamido-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
Boiling Pt: 414.4 °C (760 mm Hg)
Melting Pt: 167…168 °C
Flash Pt: 204.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00100490
CAS Number: 7298-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR