N-(3,5-Dimethyladamantan-1-yl)formamide 98%

Supplier: Ambeed
Warning

A370407-100MG A370407-250MG A370407-1G
103826-460EA 165 USD
103826-460 76989-870 76989-872 77306-810
N-(3,5-Dimethyladamantan-1-yl)formamide 98%
N-(3,5-Dimethyladamantan-1-yl)formamide

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO
MW: 207.31 g/mol
Boiling Pt: 325 °C (760 mm Hg)
Melting Pt: 69.5 °C
Storage Temperature: Freezer
MDL Number: MFCD01825787
CAS Number: 351329-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR