1-(3-Acetamidophenyl)-5-mercaptotetrazole 97%

Supplier: Ambeed
Danger

Synonyms: N-(3-(5-Mercapto-1H-tetrazol-1-yl)phenyl)acetamide

A101117-10G A101117-250MG
103795-808EA 13 USD
103795-808 103821-498 77270-198 77270-200
1-(3-Acetamidophenyl)-5-mercaptotetrazole 97%
1-(3-Acetamidophenyl)-5-mercaptotetrazole

Precaución: for Research Use Only

Formula: C₉H₉N₅OS
MW: 235.27 g/mol
Melting Pt: 201…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00603728
CAS Number: 14070-48-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR