N-(2-Oxoethyl)phthalimide 98%

Supplier: Ambeed
Warning

Synonyms: N-(Formylmethyl)phthalimide, Phthalimidoacetaldehyde

A105867-10G A105867-25G A105867-5G A105867-100G
103796-956EA 5 USD
103796-956 103822-702 76717-824 76717-826 77309-568 77878-810
N-(2-Oxoethyl)phthalimide 98%
N-(2-Oxoethyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
Melting Pt: 116…118 °C
Flash Pt: 151.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00023080
CAS Number: 2913-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR