2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine

A134137-100G A134137-10G A134137-500G A134137-5G A134137-25G
103808-478EA 316 USD
103808-478 76719-060 76719-062 76719-064 76719-066 77314-696
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine 95%
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₅O₂
MW: 307.313 g/mol
Boiling Pt: 546.6 °C (760 mm Hg)
Melting Pt: 188…193 °C
Storage Temperature: Ambient
MDL Number: MFCD02179269
CAS Number: 152460-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR