Aegeline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Aegelin, N-(2-Hydroxy-2-(4-methoxyphenyl)ethyl)cinnamamide, (E)-N-[2-Hydroxy-2-(4-methoxyphenyl)ethyl]cinnamamide
A181072-10G
A181072-5G
103807-582EA
14
USD
103807-582
103845-946
77011-998
77083-462
Aegeline 95%
Aegeline
Precaución: for Research Use Only
Formula:
C₁₈H₁₉NO₃ MW: 297.35 g/mol Boiling Pt: 567.6 °C (760 mm Hg) Melting Pt: 178…179 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00048045 CAS Number: 456-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...