N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propane-1-sulfonamide 95%

Supplier: Ambeed
Warning

A114588-100MG A114588-250MG A114588-5G
103798-426EA 326 USD
103798-426 103824-266 76646-856 76646-858 77297-312
N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propane-1-sulfonamide 95%
N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propane-1-sulfonamide

Precaución: for Research Use Only

Formula: C₁₅H₂₃BFNO₄S
MW: 343.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22494591
CAS Number: 1269233-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR