N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Bromoethylamino)propylamine dihydrobromide

A101496-100G A101496-10G A101496-250MG A101496-25G A101496-1G
103796-004EA 8 USD
103796-004 103821-688 76994-504 76994-506 76994-508 76994-510 77297-136
N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide 97%
N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide

Precaución: for Research Use Only

Formula: C₅H₁₅Br₃N₂
MW: 342.90 g/mol
Boiling Pt: 313.2 °C (760 mm Hg)
Melting Pt: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00460336
CAS Number: 23545-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR