2'-Aminoacetanilide 98%

Supplier: Ambeed
Warning

A351747-1G A351747-25G A351747-5G A351747-100G A351747-10G
103826-052EA 13 USD
103826-052 103837-048 76840-156 76840-158 76840-160 77106-796 77106-798
2'-Aminoacetanilide 98%
2'-Aminoacetanilide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 361.5 °C (760 mm Hg)
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00210388
CAS Number: 34801-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR