N-(2-aminoethyl)prop-2-enamide hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-Aminoethyl)acrylamide hydrochloride, hydrochloride, N-(2-Aminoethyl)prop-2-enamide

A153211-100MG A153211-250MG
103804-474EA 255 USD
103804-474 103842-644 77010-902 77010-904
N-(2-aminoethyl)prop-2-enamide hydrochloride 95%
N-(2-aminoethyl)prop-2-enamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O·ClH
MW: 150.607 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18380727
CAS Number: 54641-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR