N-(2,6-Difluoro-3-nitrophenyl)acetamide 95%

Supplier: Ambeed
Warning

A140048-10G A140048-250MG A140048-25G A140048-1G A140048-5G
103802-820EA 41 USD
103802-820 76719-326 76719-328 76719-330 77299-112 77299-114
N-(2,6-Difluoro-3-nitrophenyl)acetamide 95%
N-(2,6-Difluoro-3-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₆F₂N₂O₃
MW: 216.1400 g/mol
Boiling Pt: 352.7 °C (760 mm Hg)
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00174326
CAS Number: 25892-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR