N-(2-((4-Amino-3-methylphenyl)(ethyl)amino)ethyl)methanesulfonamide sulfate(2:3) 95%

Supplier: Ambeed
Danger

Synonyms: N-(2-((4-Amino-3-methylphenyl)(ethyl)amino)ethyl)methanesulphonamide sesquisulphate

A253186-10G A253186-5G A253186-1G
103812-082EA 9 USD
103812-082 103851-930 76722-642 77091-352 77304-610
N-(2-((4-Amino-3-methylphenyl)(ethyl)amino)ethyl)methanesulfonamide sulfate(2:3) 95%
N-(2-((4-Amino-3-methylphenyl)(ethyl)amino)ethyl)methanesulfonamide sulfate(2:3)

Precaución: for Research Use Only

Formula: C₂₄H₄₈N₆O₁₆S₅
Flash Pt: 229 °C
Storage Temperature: Ambient
MDL Number: MFCD00072527
CAS Number: 25646-71-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR