Tariquidar 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-((4-(2-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)phenyl)carbamoyl)-4,5-dimethoxyphenyl)quinoline-3-carboxamide

A251517-10MG A251517-1MG A251517-50MG
103791-592EA 41 USD
103791-592 103792-494 103795-098 77091-214 77091-216 77612-426
Tariquidar 98%
Tariquidar

Precaución: for Research Use Only

Formula: C₃₈H₃₈N₄O₆
MW: 646.731 g/mol
Boiling Pt: 716 °C (760 mm Hg)
Flash Pt: 386.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09837824
CAS Number: 206873-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR