N-(1-Oxoindan-5-yl)acetamide 97%

Supplier: Ambeed
Warning

A116972-100MG A116972-250MG A116972-1G A116972-5G
103798-814EA 17 USD
103798-814 103824-686 76718-364 76718-366 77311-478 77311-480
N-(1-Oxoindan-5-yl)acetamide 97%
N-(1-Oxoindan-5-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
Boiling Pt: 427.362 °C (760 mm Hg)
Melting Pt: 172 °C
Flash Pt: 194.835 °C
Storage Temperature: Ambient
MDL Number: MFCD00099465
CAS Number: 58161-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR