Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 95%

Supplier: Ambeed
Warning

Synonyms: TFFH, N-((Dimethylamino)fluoromethylene)-N-methylmethanaminium hexafluorophosphate(V)

A212162-100G A212162-10G A212162-1G A212162-5G
103818-170EA 9 USD
103818-170 103848-870 76999-638 76999-640 77303-230 77303-232
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate 95%
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₅H₁₂F₇N₂P
MW: 264.13 g/mol
Melting Pt: 104…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684443
CAS Number: 164298-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR