N-((2S)-2-Amino-2-phenylethyl)-N,N-dimethylamine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-N1,N1-Dimethyl-2-phenylethane-1,2-diamine

A206160-100MG A206160-250MG A206160-1G A206160-5G
103817-688EA 176 USD
103817-688 103848-360 77030-300 77030-302 77086-600 77086-602
N-((2S)-2-Amino-2-phenylethyl)-N,N-dimethylamine 95%
N-((2S)-2-Amino-2-phenylethyl)-N,N-dimethylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MW: 164.24 g/mol
Boiling Pt: 238.2 °C (760 mm Hg)
Flash Pt: 91.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10565843
CAS Number: 702699-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR