N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94% (Wavelength 518.0-522.0)

Supplier: Ambeed
Warning

Synonyms: N-((2-Chloro-3-((phenylimino)methyl)cyclohex-2-en-1-ylidene)methyl)aniline hydrochloride

A111657-100G A111657-25G A111657-1G A111657-5G
103797-878EA 5 USD
103797-878 103823-698 77270-964 77270-966 77284-152 77284-154
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94% (Wavelength 518.0-522.0)
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride

Precaución: for Research Use Only

Formula: C₂H₁₉ClN₂
MW: 359.29 g/mol
Melting Pt: 224 °C
Flash Pt: 240.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191759
CAS Number: 63857-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR