N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (R,R)-TsDPEN, (1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine

A325416-100G A325416-10G A325416-1G A325416-25G A325416-5G A325416-250MG
103825-402EA 8 USD
103825-402 103836-348 76802-970 76802-972 76839-286 76839-288 76839-290 77601-462
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide 97%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₂S
Melting Pt: 128…131 °C
Flash Pt: 278.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02093428
CAS Number: 144222-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR