Mupirocin lithium 95%

Supplier: Ambeed
Warning

Synonyms: Lithium 9-(((E)-4-((2S,3R,4R,5S)-3,4-dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoate

A250547-1G A250547-250MG A250547-100MG A250547-5G
103811-930EA 121 USD
103811-930 77288-996 77612-418 77685-710 77685-711
Mupirocin lithium 95%
Mupirocin lithium

Precaución: for Research Use Only

Formula: C₂₆H₄₃LiO₉
MW: 506.561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977847
CAS Number: 73346-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR