3-Methylbenzylamine 98%

Supplier: Ambeed
Danger

A739290-5G A739290-10G A739290-1G A739290-25G
103840-820EA 11 USD
103840-820 76845-052 77118-116 77118-118 77308-240 77308-242
3-Methylbenzylamine 98%
3-Methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₁N
Boiling Pt: 202…205 °C (760 mm Hg)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00008118
CAS Number: 100-81-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR