Morpholin-4-ylacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Morpholineacetic acid
A162597-100G
A162597-10G
A162597-250MG
A162597-1G
A162597-25G
A162597-5G
103805-634EA
6
USD
103805-634
103843-882
76720-170
76720-172
76720-174
77319-934
77319-936
77319-938
Morpholin-4-ylacetic acid 98%
Morpholin-4-ylacetic acid
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₃ MW: 145.160 g/mol Boiling Pt: 271.9 °C (760 mm Hg) Melting Pt: 162…164 °C Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00504633 CAS Number: 3235-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...